missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Methyl-2-pentene 97.0+%, TCI America™
$318.75 - $1015.90
Chemical Identifiers
| CAS | 674-76-0 |
|---|---|
| Molecular Formula | C6H12 |
| Molecular Weight (g/mol) | 84.16 |
| MDL Number | MFCD00065138 |
| InChI Key | LGAQJENWWYGFSN-SNAWJCMRSA-N |
| PubChem CID | 172092 |
| IUPAC Name | (2E)-4-methylpent-2-ene |
| SMILES | C\C=C\C(C)C |
Chemical Identifiers
| 674-76-0 | |
| 84.16 | |
| LGAQJENWWYGFSN-SNAWJCMRSA-N | |
| (2E)-4-methylpent-2-ene |
| C6H12 | |
| MFCD00065138 | |
| 172092 | |
| C\C=C\C(C)C |
Specifications
| 674-76-0 | |
| 58°C | |
| MFCD00065138 | |
| 2288 | |
| C\C=C\C(C)C | |
| 84.16 | |
| 84.16 | |
| Liquid |
| Colorless | |
| C6H12 | |
| 5 mL | |
| LGAQJENWWYGFSN-SNAWJCMRSA-N | |
| (2E)-4-methylpent-2-ene | |
| 172092 | |
| ≥97.0% (GC) | |
| trans-4-Methyl-2-pentene |
Safety and Handling
EINECSNumber : (2)-0031
TSCA : No