missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Hydroxymethyl)cyclohexanol 98.0+%, TCI America™

Catalog No. H12581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12581G 1 g
H12585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12581G Supplier TCI America Supplier No. H12581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3685-27-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
InChI Key VGRZISGVNOKTQU-UHFFFAOYSA-N
PubChem CID 12892866
IUPAC Name 4-(hydroxymethyl)cyclohexan-1-ol
SMILES C1CC(CCC1CO)O

Specifications

Melting Point 104°C
Color White
Quantity 1 g
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(Hydroxymethyl)cyclohexanol