missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxycyclohexanecarboxylic Acid 97.0+%, TCI America™

Catalog No. p-7130913
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11755G 5 g
H117525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11755G Supplier TCI America Supplier No. H11755G
Only null left

Chemical Identifiers

CAS 3685-26-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00077974,MFCD02093477,MFCD04038017
InChI Key HCFRWBBJISAZNK-UHFFFAOYSA-N
PubChem CID 151138
IUPAC Name 4-hydroxycyclohexane-1-carboxylic acid
SMILES OC1CCC(CC1)C(O)=O

Specifications

Melting Point 145°C
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Hydroxycyclohexanecarboxylic Acid