missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ trans-4-Hydroxy-L-proline, 99+%
Supplier: Thermo Scientific™ 121781000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
trans-4-Hydroxy-L-proline | |
0.2% (105°C, 3 hrs) max. | |
99+% | |
C5H9NO3 | |
22,191 | |
- 75.50 (20.00°C c=5,H2O) | |
trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro | |
PMMYEEVYMWASQN-DMTCNVIQSA-N | |
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid | |
131.13 | |
CHEBI:18095 | |
99+% |
51-35-4 | |
Authentic | |
Glass bottle | |
100g | |
15,4878 | |
− 75.50 | |
Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
C1C(CNC1C(=O)O)O | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
5810 | |
131.13 |
Safety and Handling
EINECSNumber : 200-091-9
RUO â Research Use Only