missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ trans-4-Hydroxy-L-proline, 99+%

Supplier:  Thermo Scientific™ 121780250

Catalog No. AC121780250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

trans-4-Hydroxy-L-proline
0.2% (105°C, 3 hrs) max.
99+%
C5H9NO3
22,191
- 75.50 (20.00°C c=5,H2O)
trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PMMYEEVYMWASQN-DMTCNVIQSA-N
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
131.13
CHEBI:18095
99+%
51-35-4
Authentic
Glass bottle
25g
15,4878
− 75.50
Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether
C1C(CNC1C(=O)O)O
95% min. (c=10, H2O, 430nm) 1cm cell
5810
131.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-091-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only