missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals trans-4-Hydroxy-L-proline, 99+%
CAS: 51-35-4 | C5H9NO3 | 131.13 g/mol
Supplier: Thermo Scientific Chemicals 121785000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| trans-4-Hydroxy-L-proline | |
| 274.0°C to 275.0°C | |
| 0.2% (105°C, 3 hrs) max. | |
| 99+% | |
| C5H9NO3 | |
| 22,191 | |
| - 75.50 (20.00°C c=5,H2O) | |
| trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro | |
| PMMYEEVYMWASQN-DMTCNVIQSA-N | |
| (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid | |
| 131.13 | |
| CHEBI:18095 | |
| 99+% |
| 51-35-4 | |
| White | |
| Authentic | |
| Glass bottle | |
| 500 g | |
| 15,4878 | |
| − 75.50 | |
| Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
| C1C(CNC1C(=O)O)O | |
| 95% min. (c=10, H2O, 430nm) 1cm cell | |
| 5810 | |
| 131.13 | |
| Crystalline Powder or Crystals |
Chemical Identifiers
| 51-35-4 | |
| 131.13 | |
| trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro | |
| CHEBI:18095 | |
| C1C(CNC1C(=O)O)O |
| C5H9NO3 | |
| PMMYEEVYMWASQN-DMTCNVIQSA-N | |
| 5810 | |
| (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid |
Safety and Handling
EINECSNumber : 200-091-9
RUO – Research Use Only