missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Decenal, 96%, may cont. ca 7% cis-isomer

Catalog No. AAL0571506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0571506 5 g
AAL0571514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0571506 Supplier Thermo Scientific Chemicals Supplier No. L0571506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 65405-70-1 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65405-70-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00065447
InChI Key CWRKZMLUDFBPAO-VOTSOKGWSA-N
Synonym trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264
PubChem CID 5702654
IUPAC Name (E)-dec-4-enal
SMILES CCCCCC=CCCC=O

Specifications

Density 0.842
Boiling Point 80°C (4 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.445
Quantity 5 g
Beilstein 4230058
Formula Weight 154.25
Percent Purity 96%
Assay may cont. ≈7% cis-isomer
Chemical Name or Material trans-4-Decenal
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 265-741-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only