missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Butylcyclohexanecarboxylic Acid 99.0+%, TCI America™

Catalog No. p-7118872
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11365G 5 g
B113625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11365G Supplier TCI America Supplier No. B11365G

Chemical Identifiers

CAS 38289-28-0
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00059560
InChI Key BALGERHMIXFENA-UHFFFAOYSA-N
Synonym trans-4-butylcyclohexanecarboxylic acid, 4-butylcyclohexanecarboxylic acid, trans-4-n-butylcyclohexanecarboxylic acid, 4-butylcyclohexane carboxylic acid, 4-n-butylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-butyl, trans-4-butylcyclohexane carboxylic acid, 4-butylcyclohexanecarboxylic acid cis-and trans-mixture, cyclohexanecarboxylic acid, 4-butyl-, trans, enamine_005334
PubChem CID 2060518
IUPAC Name 4-butylcyclohexane-1-carboxylic acid
SMILES CCCCC1CCC(CC1)C(=O)O

Specifications

Melting Point 37°C
Color White
Quantity 5 g
Formula Weight 184.28
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Butylcyclohexanecarboxylic Acid
EINECSNumber (3)-3723
TSCA No
Recommended Storage Refrigerator