missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-aminomethyl)cyclohexylamine, 97%

Catalog No. p-7041553
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6256403 1 g
AAH6256406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6256403 Supplier Thermo Scientific Chemicals Supplier No. H6256403
Only null left

CAS: 192323-07-2 | C12H24N2O2 | 228.34 g/mol

trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 192323-07-2
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD06657670,MFCD12408630,MFCD22395596
InChI Key HMMYZMWDTDJTRR-UHFFFAOYSA-N
Synonym cis-4-boc-aminomethyl cyclohexylamine, tert-butyl trans-4-aminocyclohexylmethylcarbamate, tert-butyl trans-4-aminocyclohexyl methyl carbamate, tert-butyl cis-4-aminocyclohexyl methyl carbamate, trans-4-boc-aminomethyl cyclohexylamine, 4-boc-aminomethyl-cyclohexylamine, tert-butyl n-4-aminocyclohexyl methyl carbamate, tert-butyl 4-aminocyclohexyl methyl carbamate, tert-butyl trans-4-aminocyclohexyl methylcarbamate
PubChem CID 21882540
IUPAC Name tert-butyl N-[(4-aminocyclohexyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(N)CC1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only