missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexylamine, 97%

Catalog No. AAH6273406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6273406 5 g
AAH6273414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6273406 Supplier Thermo Scientific Chemicals Supplier No. H6273406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 177906-48-8 | C11H22N2O2 | 214.31 g/mol

trans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 177906-48-8
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.31
MDL Number MFCD01076211,MFCD03001719,MFCD03844604
InChI Key FEYLUKDSKVSMSZ-UHFFFAOYSA-N
Synonym trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine
PubChem CID 2756050
IUPAC Name tert-butyl N-(4-aminocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(N)CC1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 214.3
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only