missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%

Catalog No. AAH6233803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6233803 1 g
AAH6233806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6233803 Supplier Thermo Scientific Chemicals Supplier No. H6233803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 181308-57-6 | C12H21NO3 | 227.304 g/mol

trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 181308-57-6
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD03844598
InChI Key GPDBIGSFXXKWQR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehyde
PubChem CID 11020615
IUPAC Name tert-butyl N-(4-formylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C=O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 227.3
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexanecarboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only