missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%

Catalog No. AAH6244406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6244406 5 g
AAH6244403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6244406 Supplier Thermo Scientific Chemicals Supplier No. H6244406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149423-77-8 | C14H20N2O2 | 248.33 g/mol

trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 149423-77-8
Molecular Formula C14H20N2O2
Molecular Weight (g/mol) 248.33
MDL Number MFCD03839901,MFCD06657668,MFCD07772066
InChI Key JQVBZZUMWRXDSQ-UHFFFAOYSA-N
Synonym benzyl cis-4-aminocyclohexyl carbamate, benzyl trans-4-aminocyclohexyl carbamate, n-cbz-trans-1,4-cyclohexanediamine, cis-1-n-cbz-1,4-cyclohexyldiamine, cis-n-cbz-1,4-cyclohexanediamine, 4-amino-cyclohexyl-carbamic acid benzyl ester, benzyl cis-4-aminocyclohexylcarbamate, benzyl 4-aminocyclohexyl carbamate, benzyl 1r,4r-4-aminocyclohexyl carbamate, 1-n-cbz-trans-1,4-cyclohexyldiamine
PubChem CID 18650082
IUPAC Name benzyl N-(4-aminocyclohexyl)carbamate
SMILES NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 248.32
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only