missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Aminomethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7116781
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02365G 5 g
A023625G 25 g
A0236100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A02365G Supplier TCI America Supplier No. A02365G

Chemical Identifiers

CAS 1197-18-8
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00001466
InChI Key GYDJEQRTZSCIOI-UHFFFAOYSA-N
Synonym tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
PubChem CID 5526
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CN)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(Aminomethyl)cyclohexanecarboxylic Acid
EINECSNumber (9)-0880
RTECSNumber GU8400000
TSCA No