missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(Aminomethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 157.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans-4-(Aminomethyl)cyclohexanecarboxylic Acid |
Safety and Handling
| EINECSNumber | (9)-0880 |
| RTECSNumber | GU8400000 |
| TSCA | No |