Learn More
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
CAS: 1197-18-8 | C8H15NO2 | 157.213 g/mol
$89.30 - $1006.27
Chemical Identifiers
| CAS | 1197-18-8 |
|---|---|
| Molecular Formula | C8H15NO2 |
| Molecular Weight (g/mol) | 157.213 |
| MDL Number | MFCD00001466 |
| InChI Key | GYDJEQRTZSCIOI-UHFFFAOYSA-N |
| Synonym | tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron |
| PubChem CID | 5526 |
| IUPAC Name | 4-(aminomethyl)cyclohexane-1-carboxylic acid |
| SMILES | C1CC(CCC1CN)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2174209
|
Thermo Scientific Chemicals
B2174209 |
10 g |
Each for $89.30
|
|
|||||
|
AAB2174218
|
Thermo Scientific Chemicals
B2174218 |
50 g |
Each for $295.96
|
|
|||||
|
AAB2174230
|
Thermo Scientific Chemicals
B2174230 |
250 g |
Each for $1,006.27
|
|
|||||
Description
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.
Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 1197-18-8 | |
| 157.213 | |
| GYDJEQRTZSCIOI-UHFFFAOYSA-N | |
| 5526 | |
| C1CC(CCC1CN)C(=O)O |
| C8H15NO2 | |
| MFCD00001466 | |
| tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron | |
| 4-(aminomethyl)cyclohexane-1-carboxylic acid |
Specifications
| >300°C | |
| C8H15NO2 | |
| 2207452 | |
| tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron | |
| GYDJEQRTZSCIOI-UHFFFAOYSA-N | |
| 4-(aminomethyl)cyclohexane-1-carboxylic acid | |
| 157.213 | |
| 157.21 | |
| trans-4-(Aminomethyl)cyclohexanecarboxylic acid |
| 1197-18-8 | |
| MFCD00001466 | |
| 14,9569 | |
| Soluble in water (167mg/ml),ethanol (<1mg/ml at 25°C). Very slightly soluble in ether,DMSO (<1mg/ml at 25°C),and alcohol. | |
| C1CC(CCC1CN)C(=O)O | |
| 10 g | |
| 5526 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-818-2
RTECSNumber : GU8400000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only