missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanecarboxylic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog No. p-7030638
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3111403 1 g
AAH3111406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3111403 Supplier Thermo Scientific Chemicals Supplier No. H3111403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27960-59-4 | Molecular Formula: C7H14ClNO2 | Molecular Weight: 179.644 g/mol

Used as intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 27960-59-4
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD03844587
InChI Key HXZSYUOXTKQNNY-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarboxylic acid hydrochloride, cis-4-aminocyclohexanecarboxylic acid hydrochloride, 4-aminocyclohexanecarboxylic acid hydrochloride, 4-aminocyclohexane-1-carboxylic acid hydrochloride, trans-4-aminocyclohexane carboxylic acid hydrochloride, trans-4-aminocyclohexanecarboxylic acid hcl, trans-4-aminocyclohexane-1-carboxylic acid hydrochloride, trans-4-amino-cyclohexane carboxylic acid hydrochloride, 1r,4r-4-aminocyclohexane-1-carboxylic acid hydrochloride, cis-4-aminocyclohexanecarboxylic acid hcl
PubChem CID 24720880
IUPAC Name 4-aminocyclohexane-1-carboxylic acid;hydrochloride
SMILES C1CC(CCC1C(=O)O)N.Cl

Specifications

Melting Point >300°C
Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 179.65
Percent Purity 96%
Chemical Name or Material trans-4-Aminocyclohexanecarboxylic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only