missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Octene 98.0+%, TCI America™

Catalog No. o01345ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01345ML Supplier TCI America Supplier No. O01345ML
Only null left

Chemical Identifiers

CAS 14919-01-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00039971
InChI Key YCTDZYMMFQCTEO-FNORWQNLSA-N
Synonym trans-3-octene, e-3-octene, e-oct-3-ene, 3-octene, e, 3-octene, 3e, unii-2a4p9ko860, 3-octene, 3e-3-octene, 3e-oct-3-ene, cis-3-octene
PubChem CID 638228
IUPAC Name (E)-oct-3-ene
SMILES CCCCC=CCC

Specifications

Color Colorless
Boiling Point 123°C
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-3-Octene
EINECSNumber (2)-0024
TSCA No