missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Methyl-2-pentene 99.0+%, TCI America™

Supplier:  TCI America M03125ML

 View more versions of this product

Catalog No. M03125ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

trans-3-Methyl-2-pentene
Colorless
C6H12
5 mL
trans-3-methyl-2-pentene, e-3-methyl-2-pentene, 3-methyl-trans-2-pentene, e-3-methylpent-2-ene, 2-pentene, 3-methyl-, e, 3-methylpent-2-ene, 2e-3-methylpent-2-ene, 2e-3-methyl-2-pentene #, 2-pentene, 3-methyl-, 2e, e-ch3ch=c ch3 c2h5
CCC(=CC)C
84.162
84.16
Liquid
616-12-6
70°C
MFCD00009334
2288
BEQGRRJLJLVQAQ-GQCTYLIASA-N
(E)-3-methylpent-2-ene
642661
≥99.0% (GC)

Chemical Identifiers

616-12-6
84.162
BEQGRRJLJLVQAQ-GQCTYLIASA-N
642661
CCC(=CC)C
C6H12
MFCD00009334
trans-3-methyl-2-pentene, e-3-methyl-2-pentene, 3-methyl-trans-2-pentene, e-3-methylpent-2-ene, 2-pentene, 3-methyl-, e, 3-methylpent-2-ene, 2e-3-methylpent-2-ene, 2e-3-methyl-2-pentene #, 2-pentene, 3-methyl-, 2e, e-ch3ch=c ch3 c2h5
(E)-3-methylpent-2-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0031

TSCA : No

SDS
Documents

Documents

Promotions

Promotions