missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Methyl-2-pentene 99.0+%, TCI America™

Catalog No. M03121ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03121ML 1 mL
M03125ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03121ML Supplier TCI America Supplier No. M03121ML

Chemical Identifiers

CAS 616-12-6
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009334
InChI Key BEQGRRJLJLVQAQ-GQCTYLIASA-N
Synonym trans-3-methyl-2-pentene, e-3-methyl-2-pentene, 3-methyl-trans-2-pentene, e-3-methylpent-2-ene, 2-pentene, 3-methyl-, e, 3-methylpent-2-ene, 2e-3-methylpent-2-ene, 2e-3-methyl-2-pentene #, 2-pentene, 3-methyl-, 2e, e-ch3ch=c ch3 c2h5
PubChem CID 642661
IUPAC Name (E)-3-methylpent-2-ene
SMILES CCC(=CC)C

Specifications

Color Colorless
Boiling Point 70°C
Quantity 1 mL
UN Number 2288
Formula Weight 84.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-3-Methyl-2-pentene
EINECSNumber (2)-0031
TSCA No