missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexenoic Acid 95.0+%, TCI America™

Catalog No. H007725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H007725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H007725G Supplier TCI America Supplier No. H007725G

Chemical Identifiers

CAS 1577-18-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00002786
InChI Key XXHDAWYDNSXJQM-ONEGZZNKSA-N
Synonym trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
PubChem CID 5282708
ChEBI CHEBI:49285
IUPAC Name (3E)-hex-3-enoic acid
SMILES CC\C=C\CC(O)=O

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 3265
Formula Weight 114.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-3-Hexenoic Acid
EINECSNumber (2)-0609
RTECSNumber MP7730000
TSCA Yes