missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexenoic acid, 97%

Catalog No. AAA1342809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1342809 10 g
AAA1342818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1342809 Supplier Thermo Scientific Chemicals Supplier No. A1342809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1577-18-0 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1577-18-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00002786
InChI Key XXHDAWYDNSXJQM-ONEGZZNKSA-N
Synonym trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
PubChem CID 5282708
ChEBI CHEBI:49285
IUPAC Name (E)-hex-3-enoic acid
SMILES CC\C=C\CC(O)=O

Specifications

Melting Point 11°C to 12°C
Density 0.961
Boiling Point 206°C
Flash Point 99°C (210°F)
Odor Rancid
Refractive Index 1.4398
Quantity 10 g
UN Number UN3265
Beilstein 1700862
Formula Weight 114.14
Percent Purity 97%
Chemical Name or Material trans-3-Hexenoic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-417-8
RTECSNumber MP7730000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only