missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexen-1-ol, 97%

Catalog No. AAL1054114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1054114 25 g
AAL1054106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1054114 Supplier Thermo Scientific Chemicals Supplier No. L1054114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 928-97-2 | C6H12O | 100.161 g/mol

trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 928-97-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002960
InChI Key UFLHIIWVXFIJGU-ONEGZZNKSA-N
Synonym trans-3-hexen-1-ol, e-hex-3-en-1-ol, e-3-hexen-1-ol, trans-3-hexenol, 3-hexen-1-ol, 3e-hex-3-en-1-ol, 3-hexen-1-ol, e, trans-hex-3-en-1-ol, unii-4e0nfr5b3u, 3e-hexen-1-ol
PubChem CID 5284503
IUPAC Name (E)-hex-3-en-1-ol
SMILES CCC=CCCO

Specifications

Density 0.845
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Pleasant
Refractive Index 1.439
Quantity 25 g
UN Number UN1987
Beilstein 1719713
Merck Index 14,4700
Solubility Information Slightly soluble in water.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material trans-3-Hexen-1-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 213-193-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only