missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Heptene 97.0+%, TCI America™

Catalog No. H08745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H08745G Supplier TCI America Supplier No. H08745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14686-14-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00009475
InChI Key WZHKDGJSXCTSCK-FNORWQNLSA-N
Synonym trans-3-heptene, e-hept-3-ene, e-3-heptene, 3-heptene, e, 3-heptene, 3e, unii-o4br0a824i, hept-3-ene, cis-3-heptene, 3-heptene c,t, z-hept-3-ene
PubChem CID 5357259
IUPAC Name (3E)-hept-3-ene
SMILES CCC\C=C\CC

Specifications

Color Colorless
Boiling Point 96°C
Quantity 5 g
UN Number 3295
Formula Weight 98.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-3-Heptene