missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Fluorocinnamic acid, 98%

Catalog No. p-7025378
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
AAB2081806 5 g
AAB2081814 25 g
AAB2081822 100 g
3 options

Catalog No. AAB2081806

Supplier: Thermo Scientific Chemicals B2081806

Only null left

CAS: 20595-30-6 | C9H7FO2 | 166.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20595-30-6
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD00004383
InChI Key RTSIUKMGSDOSTI-SNAWJCMRSA-N
Synonym 3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid
PubChem CID 1551219
IUPAC Name (E)-3-(3-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)F)C=CC(=O)O

Specifications

Melting Point 165°C to 168°C
Odor Odorless
Quantity 5 g
Beilstein 1365172
Formula Weight 166.15
Percent Purity 98%
Chemical Name or Material trans-3-Fluorocinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-281-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only