missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Cyanocyclohexylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog No. p-7041649
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6293803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6293803 Supplier Thermo Scientific Chemicals Supplier No. H6293803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 920966-30-9
Molecular Formula C7H13ClN2
Molecular Weight (g/mol) 160.645
MDL Number MFCD22548413
InChI Key ZBUXGIYHCTWUTA-ZJLYAJKPSA-N
Synonym trans-3-aminocyclohexanecarbonitrile hydrochloride, 1r,3r-3-aminocyclohexane-1-carbonitrile hydrochloride
PubChem CID 67278607
IUPAC Name (1R,3R)-3-aminocyclohexane-1-carbonitrile;hydrochloride
SMILES C1CC(CC(C1)N)C#N.Cl

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 160.64
Percent Purity 97%
Chemical Name or Material trans-3-Cyanocyclohexylamine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only