missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Bromocinnamic Acid 99.0+%, TCI America™

Catalog No. p-7119383
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19615G 5 g
B196125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19615G Supplier TCI America Supplier No. B19615G
Only null left

Chemical Identifiers

CAS 14473-91-7
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004382
InChI Key YEMUSDCFQUBPAL-SNAWJCMRSA-N
Synonym 3-bromocinnamic acid, 3-3-bromophenyl acrylic acid, m-bromocinnamic acid, trans-3-bromocinnamic acid, cinnamic acid, m-bromo, bw a800u, e-3-3-bromophenyl acrylic acid, 3-3-bromophenyl-2-propenoic acid, 2e-3-3-bromophenyl acrylic acid, bromocinnamic acid,3
PubChem CID 776461
IUPAC Name (2E)-3-(3-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC(Br)=C1

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 227.06
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-3-Bromocinnamic Acid
RTECSNumber GD8420000
TSCA Yes