missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Bromocinnamic acid, 98+%

Catalog No. aaa1603506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1603506 5 g
AAA1603514 25 g
AAA1603522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1603506 Supplier Thermo Scientific Chemicals Supplier No. A1603506

CAS: 14473-91-7 | C9H7BrO2 | 227.06 g/mol

trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 14473-91-7
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004382
InChI Key YEMUSDCFQUBPAL-SNAWJCMRSA-N
Synonym 3-bromocinnamic acid, 3-3-bromophenyl acrylic acid, m-bromocinnamic acid, trans-3-bromocinnamic acid, cinnamic acid, m-bromo, bw a800u, e-3-3-bromophenyl acrylic acid, 3-3-bromophenyl-2-propenoic acid, 2e-3-3-bromophenyl acrylic acid, bromocinnamic acid,3
PubChem CID 776461
IUPAC Name (E)-3-(3-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC(Br)=C1

Specifications

Melting Point 178°C to 182°C
Odor Odorless
Quantity 5 g
Beilstein 2084230
Solubility Information Insoluble in water.
Formula Weight 227.06
Percent Purity ≥98%
Chemical Name or Material trans-3-Bromocinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-267-7
RTECSNumber GD8420000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only