missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7041465
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6220703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6220703 Supplier Thermo Scientific Chemicals Supplier No. H6220703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 218772-92-0 | Molecular Formula: C12H21NO4 | Molecular Weight: 243.30 g/mol

2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 218772-92-0
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.30
MDL Number MFCD03788640,MFCD24395695
InChI Key JSGHMGKJNZTKGF-UHFFFAOYNA-N
Synonym trans-3-tert-butoxycarbonylaminocyclohexanecarboxylic acid, 1r,3r-3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, cyclohexanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-, 1r,3r-rel, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-3-aminocyclohexanecarboxylic acid, trans-3-boc-amino-cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylaminocyclohexane-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino-cyclohexanecarboxylic acid
PubChem CID 24720881
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(O)=O

Specifications

IUPAC Name 3-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Quantity 1 g
Formula Weight 243.29
Percent Purity 97%
Chemical Name or Material trans-3-(Boc-amino)cyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only