missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride, 97%

Catalog No. AAH62409MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62409MD 250 mg
AAH6240903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62409MD Supplier Thermo Scientific Chemicals Supplier No. H62409MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1222709-28-5 | C14H21ClN2O2 | 284.784 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1222709-28-5
Molecular Formula C14H21ClN2O2
Molecular Weight (g/mol) 284.784
MDL Number MFCD12404929
InChI Key SBODKRLYZILWEM-OJERSXHUSA-N
Synonym benzyl trans-3-aminocyclohexylcarbamate hydrochloride, benzyl trans-3-aminocyclohexylcarbamatehydrochloride, trans-benzyl 3-aminocyclohexylcarbamate, benzyl n-1r,3r-3-aminocyclohexyl carbamate hydrochloride
PubChem CID 71685848
IUPAC Name benzyl N-[(1R,3R)-3-aminocyclohexyl]carbamate;hydrochloride
SMILES C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)N.Cl

Specifications

Quantity 250 mg
Formula Weight 284.78
Percent Purity 97%
Chemical Name or Material trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only