Learn More
trans-3-Aminocyclohexanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
$208.30 - $208.30
Chemical Identifiers
| CAS | 862401-49-8 |
|---|---|
| Molecular Formula | C7H14ClNO2 |
| Molecular Weight (g/mol) | 179.644 |
| MDL Number | MFCD11040475 |
| InChI Key | BOUHXQPJYJANEO-KGZKBUQUSA-N |
| Synonym | trans-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hcl, 1r,3r-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hydrochloride, trans-3-aminocyclohexane carboxylic acid hydrochloride, trans-3-aminocyclohexane-1-carboxylic acid hydrochloride |
| PubChem CID | 46172878 |
| IUPAC Name | (1R,3R)-3-aminocyclohexane-1-carboxylic acid;hydrochloride |
| SMILES | C1CC(CC(C1)N)C(=O)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6223203
|
Thermo Scientific Chemicals
H6223203 |
1 g |
Each for $208.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water, glycerol. Insoluble in ethanol, chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 862401-49-8 | |
| 179.644 | |
| BOUHXQPJYJANEO-KGZKBUQUSA-N | |
| 46172878 | |
| C1CC(CC(C1)N)C(=O)O.Cl |
| C7H14ClNO2 | |
| MFCD11040475 | |
| trans-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hcl, 1r,3r-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hydrochloride, trans-3-aminocyclohexane carboxylic acid hydrochloride, trans-3-aminocyclohexane-1-carboxylic acid hydrochloride | |
| (1R,3R)-3-aminocyclohexane-1-carboxylic acid;hydrochloride |
Specifications
| 862401-49-8 | |
| MFCD11040475 | |
| Soluble in water,glycerol. Insoluble in ethanol,chloroform | |
| C1CC(CC(C1)N)C(=O)O.Cl | |
| 1 g | |
| 46172878 | |
| 97% |
| C7H14ClNO2 | |
| trans-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hcl, 1r,3r-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hydrochloride, trans-3-aminocyclohexane carboxylic acid hydrochloride, trans-3-aminocyclohexane-1-carboxylic acid hydrochloride | |
| BOUHXQPJYJANEO-KGZKBUQUSA-N | |
| (1R,3R)-3-aminocyclohexane-1-carboxylic acid;hydrochloride | |
| 179.644 | |
| 179.64 | |
| trans-3-Aminocyclohexanecarboxylic acid hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only