missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-Aminocyclohexanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water, glycerol. Insoluble in ethanol, chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Soluble in water, glycerol. Insoluble in ethanol, chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Solubility Information | Soluble in water,glycerol. Insoluble in ethanol,chloroform |
| Quantity | 1 g |
| Formula Weight | 179.64 |
| Percent Purity | 97% |
| Chemical Name or Material | trans-3-Aminocyclohexanecarboxylic acid hydrochloride |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only