missing translation for 'onlineSavingsMsg'
Learn More

trans-3,4-Dimethyl-2-pentene 98.0+%, TCI America™

Catalog No. D12565ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12565ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12565ML Supplier TCI America Supplier No. D12565ML
Only null left

Chemical Identifiers

CAS 4914-92-5
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00070566
InChI Key PPBWEVVDSRKEIK-FNORWQNLSA-N
PubChem CID 638068
IUPAC Name (E)-3,4-dimethylpent-2-ene
SMILES CC=C(C)C(C)C

Specifications

Color Colorless
Boiling Point 87°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-3,4-Dimethyl-2-pentene