missing translation for 'onlineSavingsMsg'
Learn More

trans-3,4-Difluorocinnamic Acid 98.0+%, TCI America™

Catalog No. D41611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41611G 1 g
D41615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41611G Supplier TCI America Supplier No. D41611G

Chemical Identifiers

CAS 112897-97-9
Molecular Formula C9H6F2O2
Molecular Weight (g/mol) 184.142
MDL Number MFCD00010320
InChI Key HXBOHZQZTWAEHJ-DUXPYHPUSA-N
Synonym 3,4-difluorocinnamic acid, trans-3,4-difluorocinnamic acid, e-3-3,4-difluorophenyl acrylic acid, 2e-3-3,4-difluorophenyl prop-2-enoic acid, e-3-3,4-difluorophenyl prop-2-enoic acid, 3-3,4-difluorophenyl acrylic acid, 2-propenoic acid, 3-3,4-difluorophenyl-, 2e, pubchem3107, 2-propenoic acid, 3-3,4-difluorophenyl, 3,4-difluoro cinnamic acid
PubChem CID 2733300
IUPAC Name (E)-3-(3,4-difluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)F)F

Specifications

Melting Point 196°C
Color White
Quantity 1 g
Formula Weight 184.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-3,4-Difluorocinnamic Acid