missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Undecenal 93.0+%, TCI America™

Catalog No. p-7137853
Change view
Click to view available options
Quantity:
5 mL
25 mL
Catalog No. Quantity
U00465ML 5 mL
U004625ML 25 mL
2 options

Catalog No. U00465ML

Supplier: TCI America U00465ML

Only null left

Chemical Identifiers

CAS 53448-07-0
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00014680
InChI Key PANBRUWVURLWGY-MDZDMXLPSA-N
Synonym trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30
PubChem CID 5283356
IUPAC Name (E)-undec-2-enal
SMILES CCCCCCCCC=CC=O

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 mL
Formula Weight 168.28
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Undecenal