missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenoic Acid 95.0+%, TCI America™

Catalog No. p-7134565
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03455ML 5 mL
P034525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03455ML Supplier TCI America Supplier No. P03455ML

Chemical Identifiers

CAS 13991-37-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00002704
InChI Key YIYBQIKDCADOSF-ONEGZZNKSA-N
Synonym trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
PubChem CID 638122
ChEBI CHEBI:38366
IUPAC Name (2E)-pent-2-enoic acid
SMILES CC\C=C\C(O)=O

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 mL
UN Number 3265
Formula Weight 100.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Pentenoic Acid
EINECSNumber (2)-0609
RTECSNumber SB2750000
TSCA No