missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octene 97.0+%, TCI America™

Catalog No. O01325ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01325ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01325ML Supplier TCI America Supplier No. O01325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13389-42-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (2E)-oct-2-ene
SMILES CCCCC\C=C\C

Specifications

Color Colorless
Boiling Point 125°C
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Octene
EINECSNumber (2)-0024
TSCA No