missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonenoic acid, 98%

Catalog No. AAA1322814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1322814 25 g
AAA1322822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1322814 Supplier Thermo Scientific Chemicals Supplier No. A1322814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3760-11-0 | C9H16O2 | 156.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3760-11-0
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00014020
InChI Key ADLXTJMPCFOTOO-BQYQJAHWSA-N
Synonym 2-nonenoic acid, nonylenic acid, trans-2-nonenoic acid, e-non-2-enoic acid, e-2-nonenoic acid, 2-nonenylic acid, 2e-nonenoic acid, 2-nonenoic acid, 2e, alpha-nonenoic acid, 2e-non-2-enoic acid
PubChem CID 5312586
IUPAC Name (E)-non-2-enoic acid
SMILES CCCCCCC=CC(=O)O

Specifications

Density 0.93
Boiling Point 130°C to 132°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Recognizable
Refractive Index 1.462
Quantity 25 g
UN Number UN3265
Beilstein 1721634
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material trans-2-Nonenoic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-171-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only