missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonen-1-ol 93.0+%, TCI America™

Supplier:  TCI America N058710ML

Catalog No. N058710ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

trans-2-Nonen-1-ol
Colorless
C9H18O
10 mL
CCCCCCC=CCO
142.242
142.24
Liquid
31502-14-4
104°C
MFCD00014058
NSSALFVIQPAIQK-BQYQJAHWSA-N
(E)-non-2-en-1-ol
5364941
≥93.0% (GC)

Chemical Identifiers

31502-14-4
142.242
NSSALFVIQPAIQK-BQYQJAHWSA-N
(E)-non-2-en-1-ol
C9H18O
MFCD00014058
5364941
CCCCCCC=CCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0258

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions