missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nitrocinnamic acid, 98%

Catalog No. AAA1317136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1317136 500 g
AAA1317114 25 g
AAA1317122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1317136 Supplier Thermo Scientific Chemicals Supplier No. A1317136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1013-96-3 | C9H7NO4 | 193.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1013-96-3
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007189
InChI Key BBQDLDVSEDAYAA-AATRIKPKSA-N
Synonym 2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668
PubChem CID 735923
SMILES C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point 245°C to 249°C
Odor Odorless
Quantity 500 g
Beilstein 2211209
Formula Weight 193.16
Percent Purity 98%
Chemical Name or Material trans-2-Nitrocinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-309-4
RTECSNumber UD3642850
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only