missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Methyl-2-pentenal 97.0+%, TCI America™
Supplier: TCI America M08935ML
Specifications
| trans-2-Methyl-2-pentenal | |
| Yellow | |
| C6H10O | |
| 5 mL | |
| IDEYZABHVQLHAF-GQCTYLIASA-N | |
| (E)-2-methylpent-2-enal | |
| 5319754 | |
| ≥97.0% (GC) |
| 14250-96-5 | |
| 138°C | |
| MFCD00006978 | |
| 1989 | |
| CCC=C(C)C=O | |
| 98.145 | |
| 98.15 | |
| Liquid |
Chemical Identifiers
| 14250-96-5 | |
| 98.145 | |
| IDEYZABHVQLHAF-GQCTYLIASA-N | |
| (E)-2-methylpent-2-enal |
| C6H10O | |
| MFCD00006978 | |
| 5319754 | |
| CCC=C(C)C=O |
Safety and Handling
EINECSNumber : (2)-0520
TSCA : Yes