missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexen-1-ol 95.0+%, TCI America™

$63.38 - $300.11

Chemical Identifiers

CAS 928-95-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00002927
InChI Key ZCHHRLHTBGRGOT-SNAWJCMRSA-N
Synonym trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
PubChem CID 5318042
IUPAC Name (2E)-hex-2-en-1-ol
SMILES CCC\C=C\CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H034625G
View Documents
TCI America
H034625G
25 g
Each for $63.38
Only null left
 
H0346250G
View Documents
TCI America
H0346250G
250 g
Each for $300.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

928-95-0
100.16
ZCHHRLHTBGRGOT-SNAWJCMRSA-N
5318042
CCC\C=C\CO
C6H12O
MFCD00002927
trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
(2E)-hex-2-en-1-ol

Specifications

928-95-0
162°C
MFCD00002927
trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
CCC\C=C\CO
100.16
100.16
Liquid
Colorless
C6H12O
25 g
ZCHHRLHTBGRGOT-SNAWJCMRSA-N
(2E)-hex-2-en-1-ol
5318042
≥95.0% (GC)
trans-2-Hexen-1-ol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

RTECSNumber : MP8390000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'