missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexen-1-ol 95.0+%, TCI America™

Catalog No. H034625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H034625G 25 g
H0346250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H034625G Supplier TCI America Supplier No. H034625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 928-95-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00002927
InChI Key ZCHHRLHTBGRGOT-SNAWJCMRSA-N
Synonym trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
PubChem CID 5318042
IUPAC Name (2E)-hex-2-en-1-ol
SMILES CCC\C=C\CO

Specifications

Color Colorless
Boiling Point 162°C
Quantity 25 g
Formula Weight 100.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexen-1-ol
EINECSNumber (2)-2393
RTECSNumber MP8390000
TSCA Yes