Learn More
trans-2-Hexen-1-ol, 97%
CAS: 928-95-0 | C6H12O | 100.16 g/mol
$59.49 - $631.97
Chemical Identifiers
| CAS | 928-95-0 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.16 |
| MDL Number | MFCD00002927 |
| InChI Key | ZCHHRLHTBGRGOT-SNAWJCMRSA-N |
| Synonym | trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans |
| PubChem CID | 5318042 |
| SMILES | CCC\C=C\CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1327214
|
Thermo Scientific Chemicals
A1327214 |
25 g |
Each for $59.49
|
|
|||||
|
AAA1327236
|
Thermo Scientific Chemicals
A1327236 |
500 g |
Each for $631.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 928-95-0 | |
| 100.16 | |
| ZCHHRLHTBGRGOT-SNAWJCMRSA-N | |
| 5318042 |
| C6H12O | |
| MFCD00002927 | |
| trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans | |
| CCC\C=C\CO |
Specifications
| 928-95-0 | |
| 158°C to 160°C | |
| Pleasant | |
| 1.438 | |
| 25 g | |
| 1719709 | |
| ZCHHRLHTBGRGOT-SNAWJCMRSA-N | |
| (2E)-hex-2-en-1-ol | |
| 5318042 | |
| 97% |
| 0.843 | |
| 54°C (129°F) | |
| C6H12O | |
| MFCD00002927 | |
| UN1987 | |
| trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans | |
| CCC\C=C\CO | |
| 100.16 | |
| 100.16 | |
| trans-2-Hexen-1-ol |
Safety and Handling
GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 213-191-2
RTECSNumber : MP8390000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only