missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexen-1-ol, 96%

Catalog No. AC158141000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158141000 100 mL Glass bottle
AC158140250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158141000 Supplier Thermo Scientific Chemicals Supplier No. 158141000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 928-95-0 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-95-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00002927
InChI Key ZCHHRLHTBGRGOT-SNAWJCMRSA-N
Synonym trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
PubChem CID 5318042
SMILES CCC\C=C\CO

Specifications

Color Colorless
Density 0.8440g/mL
Boiling Point 158.0°C to 160.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4410
Linear Formula CH3CH2CH2CH=CHCH2OH
Quantity 100 mL
Beilstein 01, II, 486
Specific Gravity 0.844
Solubility Information Solubility in water: slightly soluble.
IUPAC Name (2E)-hex-2-en-1-ol
Formula Weight 100.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material trans-2-Hexen-1-ol, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-191-2
RTECSNumber MP8390000
TSCA TSCA

RUO – Research Use Only