missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Dodecenol 90.0+%, TCI America™

Catalog No. D19365ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19365ML Supplier TCI America Supplier No. D19365ML
Only null left

Chemical Identifiers

CAS 69064-37-5
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00014047
InChI Key MLRYPOCSLBIUHY-ZHACJKMWSA-N
PubChem CID 5352845
IUPAC Name (E)-dodec-2-en-1-ol
SMILES CCCCCCCCCC=CCO

Specifications

Melting Point 3°C
Color Yellow
Boiling Point 141°C
Quantity 5 mL
Formula Weight 184.32
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Dodecenol
EINECSNumber (2)-0258
TSCA No