missing translation for 'onlineSavingsMsg'
Learn More

trans-2,cis-6-Nonadienal, ≥94.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019494
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019494 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019494 Supplier Merck Emd Millipore Supplier No. 05549100MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 557-48-2
Density 0.86 g/mL (at 25°C)
Flash Point 83°C
Linear Formula C2H5 CH=CHCH2 CH2 CH=CHCHO
MDL Number MFCD00007009
Boiling Point 94°C to 95°C (18 mmHg)
Synonym Violet leaf aldehyde
Type Analytical Standard
Quantity 100 mg
Formula Weight 138.21
Percent Purity ≥94.5% (GC)
Grade Analytical Standard
Show More Show Less