missing translation for 'onlineSavingsMsg'
Learn More

trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer

CAS: 557-48-2 | C9H14O | 138.21 g/mol

$64.08 - $670.82

Chemical Identifiers

CAS 557-48-2
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00007009
InChI Key HZYHMHHBBBSGHB-ODYTWBPASA-N
Synonym trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
PubChem CID 643731
ChEBI CHEBI:7610
IUPAC Name (2E,6Z)-nona-2,6-dienal
SMILES CCC=CCCC=CC=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1835603
View Documents
Thermo Scientific Chemicals
A1835603
1 g
Each for $64.08
Only null left
 
AAA1835606
View Documents
Thermo Scientific Chemicals
A1835606
5 g
Each for $137.52
Only null left
 
AAA1835614
View Documents
Thermo Scientific Chemicals
A1835614
25 g
Each for $670.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

557-48-2
138.21
HZYHMHHBBBSGHB-ODYTWBPASA-N
643731
(2E,6Z)-nona-2,6-dienal
C9H14O
MFCD00007009
trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
CHEBI:7610
CCC=CCCC=CC=O

Specifications

557-48-2
90°C (4 mmHg)
C9H14O
MFCD00007009
Air Sensitive
HZYHMHHBBBSGHB-ODYTWBPASA-N
(2E,6Z)-nona-2,6-dienal
643731
138.2
trans-2,cis-6-Nonadienal
0.866
76°C (169°F)
1.474
1 g
trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
CCC=CCCC=CC=O
138.21
CHEBI:7610
≥90%
remainder mainly trans,trans-isomer
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 209-178-6

RTECSNumber : RA5391800

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only