missing translation for 'onlineSavingsMsg'
Learn More

trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer

Catalog No. AAA1835614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1835614 25 g
AAA1835603 1 g
AAA1835606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1835614 Supplier Thermo Scientific Chemicals Supplier No. A1835614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 557-48-2 | C9H14O | 138.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 557-48-2
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00007009
InChI Key HZYHMHHBBBSGHB-ODYTWBPASA-N
Synonym trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
PubChem CID 643731
ChEBI CHEBI:7610
IUPAC Name (2E,6Z)-nona-2,6-dienal
SMILES CCC=CCCC=CC=O

Specifications

Density 0.866
Boiling Point 90°C (4 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.474
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 138.2
Percent Purity ≥90%
Assay remainder mainly trans,trans-isomer
Chemical Name or Material trans-2,cis-6-Nonadienal
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 209-178-6
RTECSNumber RA5391800
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only