missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Butenyl acetate, 95%

Catalog No. AAL0973406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0973406 5 g
AAL0973414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0973406 Supplier Thermo Scientific Chemicals Supplier No. L0973406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 628-08-0 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-08-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00039912
InChI Key WNHXJHGRIHUOTG-ONEGZZNKSA-N
Synonym trans-2-butenyl acetate, crotyl acetate, e-2-butenyl acetate, 2-buten-1-ol, acetate, e-crotyl acetate, e-but-2-enyl acetate, 2-buten-1-ol, acetate, 2e, 2-buten-1-ol, 1-acetate, 2e, 2e-but-2-en-1-yl acetate, e-2-buten-1-yl acetate
PubChem CID 5363082
IUPAC Name [(E)-but-2-enyl] acetate
SMILES CC=CCOC(=O)C

Specifications

Density 0.919
Boiling Point 128°C to 130°C
Flash Point 31°C (87°F)
Refractive Index 1.42
Quantity 5 g
UN Number UN3272
Beilstein 1720994
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material trans-2-Butenyl acetate
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only