missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Butene-1,4-diol 95.0+%, TCI America™

Catalog No. p-7122914
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33841G 1 g
B33845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33841G Supplier TCI America Supplier No. B33841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 821-11-4
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00063207
InChI Key ORTVZLZNOYNASJ-OWOJBTEDSA-N
Synonym trans-1,4-Dihydroxy-2-butene
PubChem CID 175854
IUPAC Name (E)-but-2-ene-1,4-diol
SMILES C(C=CCO)O

Specifications

Melting Point 25°C
Color White-Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 88.11
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material trans-2-Butene-1,4-diol
EINECSNumber (9)-1161
TSCA No